Skip to Content
Merck
All Photos(1)

Key Documents

8.20923

Sigma-Aldrich

cis-9-Octadecen-1-ol

for synthesis

Synonym(s):

cis-9-Octadecen-1-ol, Oleyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$42.50
1 L
$85.40
2.5 L
$184.00

$42.50


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$42.50
1 L
$85.40
2.5 L
$184.00

About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)8OH
CAS Number:
Molecular Weight:
268.48
MDL number:
UNSPSC Code:
12352104
EC Index Number:
205-597-3
NACRES:
NA.22

$42.50


Estimated to ship on30 May 2025


Request a Bulk Order

vapor pressure

10.64 hPa ( 195 °C)

Quality Level

Assay

≥80.0% (GC)

form

liquid

potency

>5000 mg/kg LD50, oral (Rat)

bp

330-360 °C/1013 hPa

mp

2-12 °C

transition temp

flash point 190 °C

density

0.85 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCCCCCCCC\C=C/CCCCCCCC

InChI

1S/C18H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h9-10,19H,2-8,11-18H2,1H3/b10-9-

InChI key

ALSTYHKOOCGGFT-KTKRTIGZSA-N

Analysis Note

Assay (GC, area%): ≥ 80.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.845 - 0.855
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

338.0 °F - closed cup

Flash Point(C)

170 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service