Skip to Content
Merck
All Photos(1)

Key Documents

8.20895

Sigma-Aldrich

3-Nitrophenol

for synthesis

Synonym(s):

3-Nitrophenol, m-Nitrophenol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$87.50
25 G
$163.00
100 G
$559.00

$87.50


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
$87.50
25 G
$163.00
100 G
$559.00

About This Item

Empirical Formula (Hill Notation):
C6H5NO3
CAS Number:
Molecular Weight:
139.11
MDL number:
UNSPSC Code:
12352104
EC Index Number:
209-073-5
NACRES:
NA.22

$87.50


Only 0 left in stock

Request a Bulk Order

Quality Level

Assay

≥99.0% (GC)

form

solid

autoignition temp.

400 °C

potency

328 mg/kg LD50, oral (Rat)

pH

6.6-8.6 (20 °C in H2O)

bp

194 °C/93 hPa

mp

95-98 °C

transition temp

flash point >100 °C

solubility

insoluble 13.5 g/L

density

1.49 g/cm3 at 20 °C

bulk density

640 kg/m3

storage temp.

2-30°C

SMILES string

[N+](=O)([O-])c1cc(ccc1)O

InChI

1S/C6H5NO3/c8-6-3-1-2-5(4-6)7(9)10/h1-4,8H

InChI key

RTZZCYNQPHTPPL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 95 °C
Melting range (upper value): ≤ 98 °C
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service