8.20873
Sodium phenolate trihydrate
for synthesis
Synonym(s):
Sodium phenolate trihydrate, Phenol sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
form
crystals
mp
56-61 °C
storage temp.
2-30°C
InChI
1S/C6H6O.Na.3H2O/c7-6-4-2-1-3-5-6;;;;/h1-5,7H;;3*1H2/q;+1;;;/p-1
InChI key
QAJGUMORHHJFNJ-UHFFFAOYSA-M
Analysis Note
Assay (HClO₄, calc. on anhydrous substance): ≥ 98.0 %
Water (K. F.): 30.0 - 33.0 %
Melting range (lower value): ≥ 56 °C
Melting range (upper value): ≤ 61 °C
Identity (IR): passes test
Water (K. F.): 30.0 - 33.0 %
Melting range (lower value): ≥ 56 °C
Melting range (upper value): ≤ 61 °C
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible, corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service