Skip to Content
Merck
All Photos(1)

Key Documents

8.20701

Sigma-Aldrich

Indene

for synthesis

Synonym(s):

Indene

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$97.60
1 L
$346.00

$97.60


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
$97.60
1 L
$346.00

About This Item

Empirical Formula (Hill Notation):
C9H8
CAS Number:
Molecular Weight:
116.16
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-393-6
NACRES:
NA.22

$97.60


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

1.7 hPa ( 20 °C)

Quality Level

Assay

≥95.0% (GC)

form

liquid

mp

-2 °C

transition temp

flash point 58 °C

density

0.99 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C1c2c(cccc2)C=C1

InChI

1S/C9H8/c1-2-5-9-7-3-6-8(9)4-1/h1-6H,7H2

InChI key

YBYIRNPNPLQARY-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.989 - 0.995
Identity (IR): passes test

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service