Skip to Content
Merck
All Photos(2)

Key Documents

8.20675

Sigma-Aldrich

N-(Hydroxymethyl)acrylamide

(48% aqueous solution) (stabilised with hydroquinone monomethyl ether) for synthesis

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$61.50
1 L
$687.00

$61.50


0 left in stock

Request a Bulk Order

Select a Size

Change View
100 ML
$61.50
1 L
$687.00

About This Item

UNSPSC Code:
12352111
NACRES:
NA.22

$61.50


0 left in stock

Request a Bulk Order

vapor pressure

31 hPa ( 25 °C)

Quality Level

form

liquid

pH

6.0-7.0 (20 °C in H2O)

bp

100 °C/1013 hPa

mp

-10 °C

transition temp

flash point >93 °C

density

1.08 g/cm3 at 20 °C

Analysis Note

Assay (ex N): 46.0 - 50.0 %
Density (d 20 °C/ 4 °C): 1.080 - 1.084
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Muta. 1B - Repr. 2 - Skin Sens. 1 - STOT RE 1 - STOT RE 2 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service