Skip to Content
Merck
All Photos(1)

Key Documents

8.20561

Sigma-Aldrich

2-Phenylacetohydrazide

for synthesis

Synonym(s):

2-Phenylacetohydrazide, Acetic acid 2-phenylhydrazide, 1-Acetyl-2-phenylhydrazine

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$165.00

$165.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
$165.00

About This Item

Empirical Formula (Hill Notation):
C8H10N2O
CAS Number:
Molecular Weight:
150.18
MDL number:
UNSPSC Code:
12352127
EC Index Number:
204-055-3
NACRES:
NA.22

$165.00


Estimated to ship on21 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

form

powder

mp

131 °C

storage temp.

15-25°C

SMILES string

N(NC(=O)C)c1ccccc1

InChI

1S/C8H10N2O/c1-7(11)9-10-8-5-3-2-4-6-8/h2-6,10H,1H3,(H,9,11)

InChI key

UICBCXONCUFSOI-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 127 °C
Melting range (upper value): ≤ 130 °C
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service