Skip to Content
Merck
All Photos(1)

Key Documents

8.20502

Sigma-Aldrich

Cumene hydroperoxide

(80% solution in cumene) for synthesis

Synonym(s):

α,α-Dimethylbenzyl hydroperoxide

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$37.50
250 ML
$69.90

$37.50


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
$37.50
250 ML
$69.90

About This Item

UNSPSC Code:
12352120
NACRES:
NA.22

$37.50


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

form

liquid

potency

573 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: oxidant

pH

>5 (20 °C, 13 g/L in H2O)

bp

116 °C/20 hPa

mp

<-30 °C

transition temp

flash point 57 °C

density

1.03 g/cm3 at 20 °C

storage temp.

15-25°C

Analysis Note

Assay (iodometric): 82.0 - 87.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Self-react. F - Skin Corr. 1B - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Lungs, Respiratory system

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 2

Flash Point(F)

134.6 °F

Flash Point(C)

57 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service