Skip to Content
Merck
All Photos(1)

Key Documents

8.20358

Sigma-Aldrich

Cyanuric acid

for synthesis

Synonym(s):

Cyanuric acid, 1,3,5-Triazine-2,4,6-triol, Isocyanuric acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$37.50
250 G
$39.70
1 KG
$109.00
50 KG
$942.00

$37.50


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
5 G
$37.50
250 G
$39.70
1 KG
$109.00
50 KG
$942.00

About This Item

Linear Formula:
C3N3(OH)3
CAS Number:
Molecular Weight:
129.07
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-618-0
NACRES:
NA.22

$37.50


Estimated to ship on01 June 2025


Request a Bulk Order

vapor pressure

<0.0001 Pa ( 25 °C)

Quality Level

form

powder

potency

7700 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

pH

3.8-4.0 (20 °C in H2O, saturated solution)

mp

320-330 °C (decomposition)

solubility

2 g/L

density

2.5 g/cm3 at 20 °C

bulk density

550 kg/m3

storage temp.

2-30°C

SMILES string

[nH]1[c]([nH][c]([nH][c]1=O)=O)=O

InChI

1S/C3H3N3O3/c7-1-4-2(8)6-3(9)5-1/h(H3,4,5,6,7,8,9)

InChI key

ZFSLODLOARCGLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. what is particle size?

    1 answer
    1. The particle size is not determined for this product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service