Skip to Content
Merck
All Photos(1)

Key Documents

8.20240

Sigma-Aldrich

tert-Butylbenzene

for synthesis

Synonym(s):

tert-Butylbenzene, 2-Methyl-2-phenylpropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H5
CAS Number:
Molecular Weight:
134.22
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-632-4
Pricing and availability is not currently available.

vapor pressure

1.33 hPa ( 13 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

445 °C

potency

3045 mg/kg LD50, oral (Rat)

expl. lim.

0.8-5.6 % (v/v)

mp

-58 °C

transition temp

flash point 44 °C

solubility

29.5 mg/L

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C10H14/c1-10(2,3)9-7-5-4-6-8-9/h4-8H,1-3H3

InChI key

YTZKOQUCBOVLHL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.865 - 0.866
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service