Skip to Content
Merck
All Photos(1)

Key Documents

8.20238

Sigma-Aldrich

4-tert-Butylbenzoic acid

for synthesis

Synonym(s):

4-tert-Butylbenzoic acid, PTBBA

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$46.10
1 KG
$88.30

$46.10


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 G
$46.10
1 KG
$88.30

About This Item

Linear Formula:
4-[(CH3)3C]C6H4COOH
CAS Number:
Molecular Weight:
178.23
MDL number:
UNSPSC Code:
12352106
EC Index Number:
202-696-3
NACRES:
NA.22

$46.10


Estimated to ship on01 June 2025


Request a Bulk Order

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (acidimetric)

form

solid

autoignition temp.

>510 °C

potency

473-735 mg/kg LD50, oral (Rat)

pH

3.9 (20 °C in H2O, saturated solution)

mp

165-167 °C

density

0.6 g/cm3 at 20 °C

bulk density

340 kg/m3

storage temp.

2-30°C

SMILES string

OC(=O)c1ccc(cc1)C(C)(C)C

InChI

1S/C11H14O2/c1-11(2,3)9-6-4-8(5-7-9)10(12)13/h4-7H,1-3H3,(H,12,13)

InChI key

KDVYCTOWXSLNNI-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Melting range (lower value): ≥ 165 °C
Melting range (upper value): ≤ 167 °C
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Repr. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service