Skip to Content
Merck
All Photos(2)

Key Documents

8.20116

Sigma-Aldrich

Azelaic acid,

technical grade for synthesis

Synonym(s):

Azelaic acid,, Nonanedioic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$50.30
1 KG
$116.00
25 KG
$1,900.00
50 KG
$3,680.00

$50.30


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
$50.30
1 KG
$116.00
25 KG
$1,900.00
50 KG
$3,680.00

About This Item

Linear Formula:
HOOC(CH2)7COOH
CAS Number:
Molecular Weight:
188.22
MDL number:
UNSPSC Code:
12352106
EC Index Number:
204-669-1
NACRES:
NA.22

$50.30


Estimated to ship on30 May 2025


Request a Bulk Order

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥90.0% (GC)

form

crystals

potency

>4000 mg/kg LD50, oral (Rat)
>10 g/kg LD50, skin (Rat)

pH

3.5 (1 g/L in H2O)

bp

237 °C/20 hPa

mp

107 °C

transition temp

flash point 215 °C

density

1.029 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[O-]C(=O)CCCCCCCC(=O)[O-].[H+].[H+]

InChI

1S/C9H16O4/c10-8(11)6-4-2-1-3-5-7-9(12)13/h1-7H2,(H,10,11)(H,12,13)

InChI key

BDJRBEYXGGNYIS-UHFFFAOYSA-N

Application

  • Enhancing Oxidative Stability in Fish Cooking Wastewater: Azelaic acid was studied for its effectiveness in treating fish cooking wastewater to enhance the oxidative stability of polyunsaturated fatty acids, demonstrating its potential in reducing environmental impact and improving wastewater treatment processes (Araújo-Rodrigues et al., 2024).
  • Comprehensive Review of Azelaic Acid: A detailed review of azelaic acid, covering its synthesis, properties, and a broad spectrum of applications, particularly highlighting its role in dermatology and industrial manufacturing processes, serving as a foundational reference for both academic research and industrial applications (2006).
  • Advancements in Hydrogel Microneedles for Cancer Treatment: Investigates the use of azelaic acid in the fabrication of hydrogel microneedles for melanoma treatment, emphasizing its role in improving delivery systems for targeted therapy and enhancing treatment efficacy (Xing et al., 2024).
  • Dermatological Applications of Azelaic Acid: Explores the diverse uses of azelaic acid in dermatology, including its mechanisms of action against acne and hyperpigmentation, and its formulation in various dermatological products, highlighting its therapeutic potential and versatility in skin treatment regimens (Sauer et al., 2023).
  • Gut Microbiota-related Metabolite Studies: Discusses azelaic acid′s role in mitigating intestinal inflammation associated with oral infections, demonstrating its importance in the study of gut microbiota-related metabolites and their potential in treating infectious diseases (Yang et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 90.0 % (a/a)
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

419.0 °F

Flash Point(C)

215 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service