Skip to Content
Merck
All Photos(1)

Key Documents

8.20054

Sigma-Aldrich

Aluminium tri-sec-butylate

for synthesis

Synonym(s):

Aluminium tri-sec-butylate, Aluminium tri-sec-butanolate, Aluminium sec-butoxide

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$53.70
1 L
$152.00

$53.70


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
$53.70
1 L
$152.00

About This Item

Linear Formula:
Al[OCH(CH3)C2H5]3
CAS Number:
Molecular Weight:
246.32
MDL number:
UNSPSC Code:
12352103
EC Index Number:
218-871-2
NACRES:
NA.22

$53.70


Estimated to ship on31 May 2025


Request a Bulk Order

vapor pressure

23 hPa ( 195 °C)

Quality Level

form

liquid

autoignition temp.

406 °C

potency

401 mg/kg LD50, oral (Rat)
402 mg/kg LD50, skin (Rabbit)

expl. lim.

1.7-9.8 % (v/v)

bp

180-182 °C/5 hPa

transition temp

flash point 26 °C

density

0.96 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Al+3].[O-]C(CC)C.[O-]C(CC)C.[O-]C(CC)C

InChI

1S/3C4H9O.Al/c3*1-3-4(2)5;/h3*4H,3H2,1-2H3;/q3*-1;+3

InChI key

WOZZOSDBXABUFO-UHFFFAOYSA-N

Application

Aluminium tri-sec-butylate can be used as:
  • A precursor in the preparation of alumina[1].
  • An initiator for the bulk ring-opening polymerization of δ-valerolactone and ε-caprolactone.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F

Flash Point(C)

26 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation of toluene in humid air by metal oxides supported on $\gamma$-alumina
Esmaeilirad M, et al.
Journal of Hazardous Materials, 333, 293-307 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service