Skip to Content
Merck
All Photos(1)

Key Documents

8.18889

Sigma-Aldrich

Hexamethyldisilane

for synthesis

Synonym(s):

Hexamethyldisilane

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$85.70
50 ML
$184.00

$85.70


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
10 ML
$85.70
50 ML
$184.00

About This Item

Linear Formula:
(CH3)3SiSi(CH3)3
CAS Number:
Molecular Weight:
146.38
MDL number:
UNSPSC Code:
12352103
EC Index Number:
215-911-0
NACRES:
NA.22

$85.70


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

2160 mg/kg LD50, oral (Rat)
2160 mg/kg LD50, skin (Rabbit)

mp

13-15 °C

transition temp

flash point -6 °C

density

0.73 g/cm3 at 20 °C

SMILES string

[Si]([Si](C)(C)C)(C)(C)C

InChI

1S/C6H18Si2/c1-7(2,3)8(4,5)6/h1-6H3

InChI key

NEXSMEBSBIABKL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.726 - 0.728
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

21.2 °F

Flash Point(C)

-6 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service