Skip to Content
Merck
All Photos(1)

Key Documents

8.18783

Sigma-Aldrich

Decanonitrile

for synthesis

Synonym(s):

Decanonitrile, Capric acid nitrile

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$768.00

$768.00


Estimated to ship on22 May 2025



Select a Size

Change View
100 ML
$768.00

About This Item

Linear Formula:
CH3(CH2)8CN
CAS Number:
Molecular Weight:
153.26
MDL number:
UNSPSC Code:
12352117
EC Index Number:
217-830-6
NACRES:
NA.22

$768.00


Estimated to ship on22 May 2025


Quality Level

Assay

≥98.0% (GC)

mp

-18 °C

transition temp

flash point 97 °C

density

0.81 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N#CCCCCCCCCC

InChI

1S/C10H19N/c1-2-3-4-5-6-7-8-9-10-11/h2-9H2,1H3

InChI key

HBZDPWBWBJMYRY-UHFFFAOYSA-N

General description

Decanonitrile, an organic compound with a nitrile group, plays a vital role in organic synthesis. The nitrile group is highly polar, making it a good electrophilic center. It can participate in nucleophilic attack, often serving as a key intermediate in the formation of other functional groups such as ketones, carboxylic acids, and amines. Additionally, decanonitrile acts as a precursor in the production of polyamides.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.818 - 0.820
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

206.6 °F - Pensky-Martens closed cup

Flash Point(C)

97 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service