Skip to Content
Merck
All Photos(1)

Key Documents

8.18775

Sigma-Aldrich

Ethyl palmitate

for synthesis

Synonym(s):

Ethyl palmitate, Palmitic acid ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$38.80
250 ML
$165.00

$38.80


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 ML
$38.80
250 ML
$165.00

About This Item

Linear Formula:
CH3(CH2)14COOC2H5
CAS Number:
Molecular Weight:
284.48
MDL number:
UNSPSC Code:
12352108
EC Index Number:
211-064-6
NACRES:
NA.22

$38.80


0 left in stock

Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

solid

bp

192-194 °C/13 hPa

mp

23-26 °C

transition temp

flash point 164 °C

density

0.86 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC)C(=O)CCCCCCCCCCCCCCC

InChI

1S/C18H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-4-2/h3-17H2,1-2H3

InChI key

XIRNKXNNONJFQO-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 21 °C
Melting range (upper value): ≤ 24 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service