Skip to Content
Merck
All Photos(2)

Documents

8.18604

Sigma-Aldrich

Tributyl phosphate

for synthesis

Synonym(s):

TBP, TBPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3(CH2)3O)3PO
CAS Number:
Molecular Weight:
266.31
Beilstein:
1710584
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-800-2
NACRES:
NA.22

vapor density

9.2 (vs air)

Quality Level

vapor pressure

27 mmHg ( 178 °C)
7.3 mmHg ( 150 °C)

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

770 °F

potency

1550 mg/kg LD50, oral (Rat)
>3100 mg/kg LD50, skin (Rabbit)

refractive index

n20/D 1.424 (lit.)

bp

180-183 °C/22 mmHg (lit.)

mp

−79 °C (lit.)

transition temp

flash point 146 °C

solubility

6 g/L

density

0.979 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CCCCOP(=O)(OCCCC)OCCCC

InChI

1S/C12H27O4P/c1-4-7-10-14-17(13,15-11-8-5-2)16-12-9-6-3/h4-12H2,1-3H3

InChI key

STCOOQWBFONSKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Spontaneous Surface Convection and Extraction (Stripping) Rate in Systems with Tributyl Phosphate and Di(2-ethylhexyl) Hydrogen Phosphate.: This study investigates the spontaneous surface convection and extraction rates in systems involving tributyl phosphate (TBP) and di(2-ethylhexyl) hydrogen phosphate. The findings are crucial for improving the efficiency of chemical separations and extractions in analytical chemistry (Kizim and Golubina, 2020).

  • The effect of radiolysis and thermally stimulated acid hydrolysis on tributyl phosphate and its solutions in ISOPAR-M.: This research explores the effects of radiolysis and thermally stimulated acid hydrolysis on TBP and its solutions in ISOPAR-M. Understanding these effects is essential for ensuring the stability and safety of TBP in various industrial applications, particularly in nuclear fuel reprocessing (Serenko et al., 2022).

  • Protective Effects in the Radiolysis of Acidified Hydrocarbon Solutions of Tributyl Phosphate.: This paper examines the protective effects in the radiolysis of acidified hydrocarbon solutions of TBP. The study provides insights into mitigating the degradation of TBP under radiation, which is significant for its application in the nuclear industry (Serenko et al., 2022).

  • Radicals from tributyl phosphate decomposition: a combined electron paramagnetic resonance spectroscopic and computational chemistry investigation.: Utilizing electron paramagnetic resonance (EPR) spectroscopy and computational chemistry, this study investigates the radicals formed from TBP decomposition. The results enhance the understanding of TBP′s stability and decomposition pathways under various conditions (Sosulin et al., 2023).

  • Kinetics and mechanism of Eu(III) transfer in tributyl phosphate microdroplet/HNO3 aqueous solution system revealed by fluorescence microspectroscopy.: This research employs fluorescence microspectroscopy to reveal the kinetics and mechanism of Europium (III) transfer in a TBP microdroplet/HNO3 system. The findings contribute to the development of more efficient separation processes in analytical and nuclear chemistry (Miyagawa et al., 2022).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.976 - 0.978
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

294.8 °F

Flash Point(C)

146 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service