Skip to Content
Merck
All Photos(1)

Key Documents

8.18559

Sigma-Aldrich

Allyl cyanoacetate

for synthesis

Synonym(s):

Allyl cyanoacetate, Cyanoacetic acid allyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$352.00
1 L
$5,650.00

$352.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
50 ML
$352.00
1 L
$5,650.00

About This Item

Linear Formula:
NCCH2COOCH2CH=CH2
CAS Number:
Molecular Weight:
125.13
MDL number:
UNSPSC Code:
12352117
EC Index Number:
236-423-4
NACRES:
NA.22

$352.00


Estimated to ship on23 May 2025


Request a Bulk Order

vapor pressure

0.05 hPa

Quality Level

Assay

≥99.0% (GC)

form

liquid

potency

160 mg/kg LD50, oral (Rat)

bp

89-90 °C/5 hPa

mp

-40.0 °C

transition temp

flash point 91 °C

density

1.072 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N#CCC(=O)OCC=C

InChI

1S/C6H7NO2/c1-2-5-9-6(8)3-4-7/h2H,1,3,5H2

InChI key

WXKCRCGKCOKJEF-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.072 - 1.073
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service