Skip to Content
Merck
All Photos(1)

Key Documents

8.18553

Sigma-Aldrich

Nerolidol

(mixture of cis- and trans-isomers) for synthesis

Synonym(s):

Nerolidol, 3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$136.00

$136.00


Estimated to ship on01 June 2025



Select a Size

Change View
100 ML
$136.00

About This Item

Linear Formula:
(CH3)2C[=CHCH2CH2C(CH3)]2(OH)CH=CH2
CAS Number:
Molecular Weight:
222.37
MDL number:
UNSPSC Code:
12352104
EC Index Number:
230-597-5
NACRES:
NA.22

$136.00


Estimated to ship on01 June 2025


vapor pressure

<1 hPa ( 20 °C)

Quality Level

autoignition temp.

230 °C

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.7-4.5 % (v/v)

bp

263-266 °C/1013 hPa

mp

-75 °C

transition temp

flash point 96 °C

solubility

0.014 g/L

density

0.88 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(CC\C=C(\CCC=C(C)C)/C)(C)C=C

InChI

1S/C15H26O/c1-6-15(5,16)12-8-11-14(4)10-7-9-13(2)3/h6,9,11,16H,1,7-8,10,12H2,2-5H3/b14-11+

InChI key

FQTLCLSUCSAZDY-SDNWHVSQSA-N

Analysis Note

Assay isomers (GC, area%): ≥ 95.0 % (a/a)
cis-Nerolidol (GC, area%): 35.0 - 45.0 % (a/a)
trans-Nerolidol (GC, area%): 50.0 - 60.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.870 - 0.880
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

262.4 °F - closed cup

Flash Point(C)

128 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service