Skip to Content
Merck
All Photos(1)

Key Documents

8.18490

Sigma-Aldrich

Methyl 4-formylbenzoate

for synthesis

Synonym(s):

Methyl 4-formylbenzoate, 4-Formylbenzoic acid methyl ester, Terephthalaldehydic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$136.00

$136.00


Estimated to ship on26 March 2025


Request a Bulk Order

Select a Size

Change View
100 G
$136.00

About This Item

Linear Formula:
4-(OHC)C6H4COOCH3
CAS Number:
Molecular Weight:
164.16
MDL number:
UNSPSC Code:
12352108
EC Index Number:
216-385-5
NACRES:
NA.22

$136.00


Estimated to ship on26 March 2025


Request a Bulk Order

Quality Level

Assay

≥96.0% (GC)

form

powder

autoignition temp.

240 °C

bp

264-266 °C/1013 hPa

mp

56-59 °C

transition temp

flash point 128 °C

density

1.2 g/cm3 at 20 °C

bulk density

528 kg/m3

storage temp.

2-30°C

SMILES string

O(C)C(=O)c1ccc(cc1)C=O

InChI

1S/C9H8O3/c1-12-9(11)8-4-2-7(6-10)3-5-8/h2-6H,1H3

InChI key

FEIOASZZURHTHB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Melting range (lower value): ≥ 60 °C
Melting range (upper value): ≤ 63 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

262.4 °F

Flash Point(C)

128 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service