Skip to Content
Merck
All Photos(2)

Key Documents

8.18405

Sigma-Aldrich

(1S)-(-)-α-Pinene

for synthesis

Synonym(s):

(1S)-(-)-α-Pinene, (1S)-2,6,6-Trimethylbicyclo[3.1.1]hept-2-ene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$38.30
500 ML
$98.70

$38.30


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$38.30
500 ML
$98.70

About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
MDL number:
UNSPSC Code:
12352002
EC Index Number:
232-077-3
NACRES:
NA.22

$38.30


Estimated to ship on31 May 2025


Request a Bulk Order

vapor pressure

5 hPa ( 25 °C)

Quality Level

Assay

≥95.0% (GC)

form

liquid

autoignition temp.

255 °C

expl. lim.

0.8 % (v/v)

reaction suitability

reagent type: reductant

mp

-62 °C

transition temp

flash point 32 °C

density

0.86 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

[C@@H]21C([C@@H](C2)C(=CC1)C)(C)C

InChI

1S/C10H16/c1-7-4-5-8-6-9(7)10(8,2)3/h4,8-9H,5-6H2,1-3H3/t8-,9-/m0/s1

InChI key

GRWFGVWFFZKLTI-IUCAKERBSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)

Density (d 20 °C/ 4 °C): 0.855 - 0.860

Spec. rotation [α²0/D (undiluted): -50.0 --20.0 °

Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

88.3 °F

Flash Point(C)

31.3 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service