Skip to Content
Merck
All Photos(1)

Key Documents

8.18369

Sigma-Aldrich

Semicarbazide hydrochloride

for synthesis

Synonym(s):

Semicarbazide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$79.10
500 G
$320.00

$79.10


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
$79.10
500 G
$320.00

About This Item

Empirical Formula (Hill Notation):
CH5N3O · xHCl
CAS Number:
Molecular Weight:
75.07 (free base basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
209-247-0
NACRES:
NA.22

$79.10


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

form

solid

potency

123 mg/kg LD50, oral (Rat)

pH

1.5 (20 °C, 100 g/L in H2O)

mp

175-177 °C (decomposition)

bulk density

740 kg/m3

storage temp.

15-25°C

SMILES string

[Cl-].N(N)C(=O)N.[H+]

InChI

1S/CH5N3O.ClH/c2-1(5)4-3;/h3H2,(H3,2,4,5);1H

InChI key

XHQYBDSXTDXSHY-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 98.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Bone

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service