Skip to Content
Merck
All Photos(1)

Key Documents

8.18313

Sigma-Aldrich

tert-Butyldimethylsilyl trifluoromethanesulfonate

for synthesis

Synonym(s):

tert-Butyldimethylsilyl trifluoromethanesulfonate, Trifluoromethanesulfonic acid tert-butyldimethylsilyl ester, tert-Butyldimethylsilyl triflate

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$527.00

$527.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
25 ML
$527.00

About This Item

Linear Formula:
CF3SO3Si(CH3)2C(CH3)3
CAS Number:
Molecular Weight:
264.34
MDL number:
UNSPSC Code:
12352103
EC Index Number:
274-102-0
NACRES:
NA.22

$527.00


Estimated to ship on21 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (acidimetric)

form

liquid

bp

65-67 °C/13 hPa

transition temp

flash point 36 °C

density

1.151 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

[Si](O[S](=O)(=O)C(F)(F)F)(C(C)(C)C)(C)C

InChI

1S/C7H15F3O3SSi/c1-6(2,3)15(4,5)13-14(11,12)7(8,9)10/h1-5H3

InChI key

WLLIXJBWWFGEHT-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 1.145 - 1.152
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service