Skip to Content
Merck
All Photos(2)

Key Documents

8.18284

Sigma-Aldrich

2′,5′-Dihydroxyacetophenone

for synthesis

Synonym(s):

2′,5′-Dihydroxyacetophenone

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$108.00

$108.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
$108.00

About This Item

Linear Formula:
2,5-(OH)2-C6H3COCH3
CAS Number:
Molecular Weight:
152.15
MDL number:
UNSPSC Code:
12352115
EC Index Number:
207-716-4
NACRES:
NA.22

$108.00


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

form

powder

mp

199-202 °C

storage temp.

2-30°C

SMILES string

Oc1c(cc(cc1)O)C(=O)C

InChI

1S/C8H8O3/c1-5(9)7-4-6(10)2-3-8(7)11/h2-4,10-11H,1H3

InChI key

WLDWSGZHNBANIO-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 200 °C
Melting range (upper value): ≤ 203 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service