Skip to Content
Merck
All Photos(2)

Key Documents

8.18204

Sigma-Aldrich

N-(Trimethylsilyl)imidazole

for synthesis

Synonym(s):

N-(Trimethylsilyl)imidazole, TSIM

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$104.00
100 ML
$322.00

$104.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
25 ML
$104.00
100 ML
$322.00

About This Item

Empirical Formula (Hill Notation):
C6H12N2Si
CAS Number:
Molecular Weight:
140.26
MDL number:
UNSPSC Code:
12352103
EC Index Number:
242-040-3
NACRES:
NA.22

$104.00


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

form

liquid

autoignition temp.

540 °C (DIN 51794)

kinematic viscosity

2.5 cSt(25 °C)

bp

212 °C (decomposition)

mp

-42 °C

transition temp

flash point 66 °C (DIN 51755 Part 1)

density

0.955-0.959 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

[Si]([n]1cncc1)(C)(C)C

InChI

1S/C6H12N2Si/c1-9(2,3)8-5-4-7-6-8/h4-6H,1-3H3

InChI key

YKFRUJSEPGHZFJ-UHFFFAOYSA-N

Application


  • Sterol Analysis: Describes the use of N-trimethylsilylimidazole for derivatizing sterols to facilitate fast GLC/MS analysis, which can be completed in under 12.5 minutes. This application is significant for analytical chemistry, particularly in sterol analysis (M Giera, F Plossl, F Bracher, 2023).

  • Synthesis of Aryl-5-arylideneimidazol-4-ones: Discusses a new method using N-Trimethylsilylimidazole for the synthesis of 2,3-diaryl-5-arylidene-4imidazolones, involving a dehydration reaction of N-substituted α,β-unsaturated carbonyl compounds, which is pertinent to medicinal chemistry (VO Topuzyan, SR Tosunyan, ER Aleksanyan, 2023).

  • Preparation of Ionic Liquids: Outlines the synthesis of 1, 3-bis-(3-sulfonic acid) propyl imidazolium ionic liquids using N-trimethylsilylimidazole, illustrating its utility in the development of new ionic liquid frameworks useful in materials science (K Yuan, W Huang, T Zhang, L Lv, Y Wang, Z Zou, 2023).

  • Ultrasonic synthesis of stable oil filled microcapsules: This study explores the ultrasonic synthesis of oil-filled microcapsules using thiolated chitosan, potentially involving derivatives like ᴅʟ-N-Acetylhomocysteine thiolactone for improved stability and functional properties. The research provides insights into advanced microencapsulation techniques relevant in pharmaceutical and industrial applications (Source).

Analysis Note

Assay (HClO₄): ≥ 97.0 %
Density (d 20 °C/ 4 °C): 0.952 - 0.958
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service