Skip to Content
Merck
All Photos(1)

Key Documents

8.18079

Sigma-Aldrich

Methyltrialkylammonium chloride

(mixture of C8-C10) for synthesis

Synonym(s):

Methyltrialkylammonium chloride, Trioctylmethylammonium chloride, Aliquat®* (* = Trade mark of Henkel Corp. USA), Methyltrioctylamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$43.10
250 ML
$60.90
1 L
$163.00

$43.10


Estimated to ship on01 April 2025


Request a Bulk Order

Select a Size

Change View
5 ML
$43.10
250 ML
$60.90
1 L
$163.00

About This Item

UNSPSC Code:
12352107
EC Index Number:
264-120-7
NACRES:
NA.22

$43.10


Estimated to ship on01 April 2025


Request a Bulk Order

vapor pressure

<1.3 hPa

Quality Level

form

liquid

potency

220 mg/kg LD50, oral (Rat)

bp

>240 °C/1013 hPa

transition temp

flash point 113 °C

density

0.89 g/cm3 at 20 °C

storage temp.

2-30°C

Application

  • Cu-CATALYZED “ON-WATER” REARRANGEMENT OF 2-AMINOBENZOTHIAZOLES TO PHENOTHIAZINES: Research on a novel “on-water” catalytic system utilizing Methyltrialkylammonium chloride for the rearrangement of 2-aminobenzothiazoles (Abele & Abele).
  • IL-Functionalized Mg3Al-LDH as New Efficient Adsorbent for Pd Recovery from Aqueous Solutions: This research focuses on the functionalization of Mg3Al-LDH with Methyltrialkylammonium chloride for efficient palladium recovery (Cocheci et al., 2022).
  • Calixresorcin arene-Mediated Transport of Pb (II) Ions through Polymer Inclusion Membrane: Discusses the use of Methyltrialkylammonium chloride in enhancing the transport of lead ions through polymer inclusion membranes (Konczyk & Ciesielski, 2021).

Analysis Note

Assay (argentometric): 85.0 - 95.0 %
Density (d 20 °C/ 4 °C): 0.885 - 0.891
Water (K. F.): 2.0 - 5.0 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Legal Information

Aliquat is a registered trademark of BASF

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C - STOT RE 2 Oral

Target Organs

Heart

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service