Skip to Content
Merck
All Photos(1)

Key Documents

8.18053

Sigma-Aldrich

Sodium cyanoborohydride

for synthesis

Synonym(s):

Sodium cyanoborohydride, Sodium cyanotrihydridoborate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$102.00
10 G
$104.00
25 G
$187.00
250 G
$1,180.00
500 G
$2,230.00

$102.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
$102.00
10 G
$104.00
25 G
$187.00
250 G
$1,180.00
500 G
$2,230.00

About This Item

Empirical Formula (Hill Notation):
CH3BNNa
CAS Number:
Molecular Weight:
62.84
MDL number:
UNSPSC Code:
12352306
EC Index Number:
247-317-2
NACRES:
NA.22

$102.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

form

powder

reaction suitability

reagent type: reductant

mp

241-243 °C

transition temp

flash point 70 °C

density

1.12 g/cm3 at 28 °C

storage temp.

2-30°C

SMILES string

[Na+].[B+3].N#[C-].[H-].[H-].[H-]

InChI

1S/CN.B.Na.3H/c1-2;;;;;/q-1;+3;+1;3*-1

InChI key

IVGYREMSCQFSBM-UHFFFAOYSA-N

Application


  • Protocols for Efficient Chemical Synthesis of N2‐Modified Guanosine Phosphate Derivatives: This research describes methods using sodium cyanoborohydride in the chemical synthesis of guanosine phosphate derivatives, essential for mRNA labeling in drug discovery applications (A Senthilvelan et al., 2023).

  • Synergistic Effect of Sodium Cyanoborohydride in Pb‐Sn Perovskite Solar Cells: Discusses the use of sodium cyanoborohydride as a multifunctional reducing agent at interfaces in perovskite solar cells, pertinent to material science, particularly in energy-related applications (J Song et al., 2023).

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials, which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

158.0 °F

Flash Point(C)

70 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service