Skip to Content
Merck
All Photos(1)

Key Documents

8.16146

Sigma-Aldrich

1-Methylpyrrolidine

for synthesis

Synonym(s):

1-Methylpyrrolidine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$59.40
10 ML
$62.90

$59.40


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$59.40
10 ML
$62.90

About This Item

Empirical Formula (Hill Notation):
C5H11N
CAS Number:
Molecular Weight:
85.15
MDL number:
UNSPSC Code:
12352005
EC Index Number:
204-438-5
NACRES:
NA.22

$59.40


Estimated to ship on25 May 2025


Request a Bulk Order

vapor pressure

100 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

170 °C (DIN 51794)

potency

280 mg/kg LD50, oral (Rat)

expl. lim.

1.1-9.1 % (v/v)

pH

11.3 (1 g/L in H2O)

bp

81 °C/1013 hPa

mp

<-80 °C

transition temp

flash point -3 °C

solubility

213 g/L

density

0.799 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N1(CCCC1)C

InChI

1S/C5H11N/c1-6-4-2-3-5-6/h2-5H2,1H3

InChI key

AVFZOVWCLRSYKC-UHFFFAOYSA-N

General description

1-Methylpyrrolidine is a heterocyclic organic compound that serves as a base and catalyst in organic reactions, such as aldol condensations, acylations, and cyclization reactions. Additionally, it is used as a solvent for certain polymerization processes due to its ability to dissolve both polar and non-polar substances. It exhibits strong nucleophilic properties because of the lone pair of electrons on its nitrogen atom, making it highly reactive towards electrophiles. This property enables its use in alkylation, acylation, and substitution reactions, facilitating the synthesis of diverse nitrogen-containing compounds.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service