Skip to Content
Merck
All Photos(1)

Key Documents

8.16102

Sigma-Aldrich

Octyl gallate

for synthesis

Synonym(s):

Octyl gallate, Gallic acid octyl ester, Octyl gallate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O5
CAS Number:
Molecular Weight:
282.33
MDL number:
UNSPSC Code:
12352108
EC Index Number:
213-853-0
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (HPLC)

potency

1960 mg/kg LD50, oral (Rat)

mp

100-101 °C

InChI

1S/C15H22O5/c1-2-3-4-5-6-7-8-20-15(19)11-9-12(16)14(18)13(17)10-11/h9-10,16-18H,2-8H2,1H3

InChI key

NRPKURNSADTHLJ-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 98 °C
Melting range (upper value): ≤ 102 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service