Skip to Content
Merck
All Photos(2)

Key Documents

8.16017

Sigma-Aldrich

(S)-(-)-Tryptophan

for synthesis

Synonym(s):

(S)-(-)-Tryptophan, Trp

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$67.50
100 G
$231.00

$67.50


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
$67.50
100 G
$231.00

About This Item

Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-795-6
NACRES:
NA.22

$67.50


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

form

solid

potency

16000 mg/kg LD50, oral (Rat)

pH

5.5-7.0 (20 °C, 10 g/L in H2O)

mp

290 °C (decomposition)

solubility

10 g/L

bulk density

400 kg/m3

storage temp.

2-30°C

SMILES string

[nH]1c2c(c(c1)CC(N)C(=O)O)cccc2

InChI

1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)

InChI key

QIVBCDIJIAJPQS-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Spec. rotation [α²0/D (c=1 in water): -32.5 - -30.5 °
Identity (IR): passes test

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service