8.14993
3-Methylbutyl isovalerate
for synthesis
Synonym(s):
3-Methylbutyl isovalerate, Isopentyl isopentanoate, Isoamyl isovalerate
Select a Size
$103.00
Estimated to ship on29 May 2025
Select a Size
About This Item
$103.00
Estimated to ship on29 May 2025
Recommended Products
vapor pressure
0.8 hPa ( 20 °C)
Quality Level
potency
>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)
bp
195 °C/1013 hPa
transition temp
flash point 80 °C
solubility
0.016 g/L
density
0.855 g/cm3 at 25 °C
storage temp.
2-30°C
SMILES string
O(CCC(C)C)C(=O)CC(C)C
InChI
1S/C10H20O2/c1-8(2)5-6-12-10(11)7-9(3)4/h8-9H,5-7H2,1-4H3
InChI key
XINCECQTMHSORG-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 0.852 - 0.857
Identity (IR): passes test
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
176.0 °F
Flash Point(C)
80 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service