Skip to Content
Merck
All Photos(1)

Key Documents

8.14992

Sigma-Aldrich

Hexyl butyrate

for synthesis

Synonym(s):

Hexyl butyrate, Butyric acid hexyl ester, Hexyl butanoate

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$227.00

$227.00


Estimated to ship on18 May 2025



Select a Size

Change View
250 ML
$227.00

About This Item

Empirical Formula (Hill Notation):
C10H20O2
CAS Number:
Molecular Weight:
172.26
MDL number:
UNSPSC Code:
12352108
EC Index Number:
220-136-6
NACRES:
NA.22

$227.00


Estimated to ship on18 May 2025


Quality Level

Assay

≥98.0% (GC)

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

mp

-78 °C

transition temp

flash point 83 °C

density

0.865 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCCCCC)C(=O)CCC

InChI

1S/C10H20O2/c1-3-5-6-7-9-12-10(11)8-4-2/h3-9H2,1-2H3

InChI key

XAPCMTMQBXLDBB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.862 - 0.867
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service