Skip to Content
Merck
All Photos(1)

Documents

8.14874

Sigma-Aldrich

2-Methyl-2-butene

(mixture of isomers) for synthesis

Synonym(s):

2-Methyl-2-butene, Isoamylene, Trimethylethylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10
CAS Number:
Molecular Weight:
70.13
UNSPSC Code:
12352002
EC Index Number:
247-975-0
NACRES:
NA.22

vapor pressure

513 hPa ( 20 °C)

Quality Level

Assay

83.0-88.0% (GC)

form

liquid

autoignition temp.

290 °C

potency

1000-1700 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

expl. lim.

1-9 % (v/v)

kinematic viscosity

0.29 cSt(20 °C)

mp

-134 °C

transition temp

flash point -45 °C

density

0.66 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/C5H10/c1-4-5(2)3/h2,4H2,1,3H3

InChI key

MHNNAWXXUZQSNM-UHFFFAOYSA-N

Application

  • Palladium-Catalyzed Pentenylation of Indoles: A study demonstrated the use of 2-Methyl-2-butene in palladium-catalyzed intermolecular dehydrogenative pentenylation of indoles, offering a novel pathway in synthetic chemistry for creating complex molecules (Zhang et al., 2024).
  • Asynchronous Concerted Mechanism: Research explored an asynchronous concerted mechanism in Lewis acid-mediated carbonyl-olefin [2+2] cycloaddition involving 2-Methyl-2-butene, providing insights into reaction dynamics and stereochemical outcomes in organic synthesis (Li et al., 2023).

Analysis Note

Assay (GC, area%): 83.0 - 88.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.658 - 0.663
2-Methyl-1-buten (GC, area%): 12.0 - 17.0 % (a/a)
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 1 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-49.0 °F

Flash Point(C)

-45 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service