Skip to Content
Merck
All Photos(1)

Key Documents

8.14692

Sigma-Aldrich

Hexadecyltrimethylammonium-p-toluenesulfonate

for synthesis

Synonym(s):

Hexadecyltrimethylammonium-p-toluenesulfonate, Cetyltrimethylammonium tosylate, Cetats, Tricetol

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$139.00

$139.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
$139.00

About This Item

Empirical Formula (Hill Notation):
C26H49NO3S
CAS Number:
Molecular Weight:
455.74
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-324-8
NACRES:
NA.22

$139.00


Estimated to ship on20 May 2025


Request a Bulk Order

Quality Level

Assay

≥99.0% (HPLC)

form

powder

potency

675 mg/kg LD50, oral (Rat)

pH

6-8 (20 °C in H2O, saturated aqueous solution)

mp

245-250 °C

solubility

10 g/L

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(O)c1ccc(cc1)C.[N+](CCCCCCCCCCCCCCCC)(C)(C)C

InChI

1S/C19H42N.C7H8O3S/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;1-6-2-4-7(5-3-6)11(8,9)10/h5-19H2,1-4H3;2-5H,1H3,(H,8,9,10)/q+1;

InChI key

MZMRZONIDDFOGF-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 245 °C
Melting range (upper value): ≤ 255 °C
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service