Skip to Content
Merck
All Photos(1)

Key Documents

8.14659

Sigma-Aldrich

Dimethyl glutarate

for synthesis

Synonym(s):

Dimethyl glutarate, Pentanedioic acid dimethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$53.80
1 L
$135.00

$53.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$53.80
1 L
$135.00

About This Item

Linear Formula:
CH3-OOC-(CH2)3-COO-CH3
CAS Number:
Molecular Weight:
160.17
MDL number:
UNSPSC Code:
12352108
EC Index Number:
214-277-2
NACRES:
NA.22

$53.80


Check Cart for Availability

Request a Bulk Order

vapor pressure

0.13 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

365 °C

potency

8191 mg/kg LD50, oral (Rat)
>2250 mg/kg LD50, skin (Rabbit)

expl. lim.

0.9-7.9 % (v/v)

kinematic viscosity

2.5 cSt(25 °C)

bp

210-215 °C/1013 hPa

mp

-37 °C

transition temp

flash point 107 °C

density

1.09 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)C(=O)CCCC(=O)OC

InChI

1S/C7H12O4/c1-10-6(8)4-3-5-7(9)11-2/h3-5H2,1-2H3

InChI key

XTDYIOOONNVFMA-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service