Skip to Content
Merck
All Photos(1)

Key Documents

8.14653

Sigma-Aldrich

Formamidine acetate

for synthesis

Synonym(s):

Formamidine acetate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$137.00
500 G
$355.00

$137.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 G
$137.00
500 G
$355.00

About This Item

Empirical Formula (Hill Notation):
CH4N2 · xC2H4O2
CAS Number:
Molecular Weight:
44.06 (free base basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
222-442-5
NACRES:
NA.22

$137.00


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

form

crystals

autoignition temp.

565 °C

potency

2510 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

pH

8 (20 °C, 400 g/L in H2O)

mp

156-160 °C

solubility

832 g/L

storage temp.

2-30°C

SMILES string

[N+H3]C=N.[O-]C(=O)C

InChI

1S/C2H4O2.CH4N2/c1-2(3)4;2-1-3/h1H3,(H,3,4);1H,(H3,2,3)

InChI key

XPOLVIIHTDKJRY-UHFFFAOYSA-N

Analysis Note

Color (visual): colourless to yellowish powder
Assay (HClO₄): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service