Skip to Content
Merck
All Photos(1)

Key Documents

8.14651

Sigma-Aldrich

Tetramethyl orthosilicate

for synthesis

Synonym(s):

Tetramethyl orthosilicate

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$85.20

$85.20


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
25 ML
$85.20

About This Item

Empirical Formula (Hill Notation):
C4H12O4Si
CAS Number:
Molecular Weight:
152.22
MDL number:
UNSPSC Code:
12352103
EC Index Number:
211-656-4
NACRES:
NA.22

$85.20


Only 0 left in stock

Request a Bulk Order

vapor pressure

18 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

245 °C

potency

17510 mg/kg LD50, skin (Rabbit)

expl. lim.

0.88-23.8 % (v/v)

bp

122 °C/1013 hPa

mp

3 °C

transition temp

flash point 26 °C

density

1.03 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](OC)(OC)(OC)OC

InChI

1S/C4H12O4Si/c1-5-9(6-2,7-3)8-4/h1-4H3

InChI key

LFQCEHFDDXELDD-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.030 - 1.035
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service