Skip to Content
Merck
All Photos(1)

Key Documents

8.14637

Sigma-Aldrich

Methanesulfonic acid

(70% solution in water) for synthesis

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$55.30
1 L
$174.00

$55.30


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
$55.30
1 L
$174.00

About This Item

UNSPSC Code:
12352106
NACRES:
NA.22

$55.30


Estimated to ship on31 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

form

liquid

potency

1158 mg/kg LD50, oral (Rat)

mp

-60 °C

density

1.35 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay (acidimetric): 69.0 - 71.0 %
Density (d 20 °C/ 4 °C): 1.350 - 1.360

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service