Skip to Content
Merck
All Photos(1)

Key Documents

8.14549

Sigma-Aldrich

Formaldehyde diethyl acetal

for synthesis

Synonym(s):

Formaldehyde diethyl acetal, Diethoxymethane

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$47.80
1 L
$94.30

$47.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$47.80
1 L
$94.30

About This Item

Empirical Formula (Hill Notation):
C5H12O2
CAS Number:
Molecular Weight:
104.15
MDL number:
UNSPSC Code:
12352112
EC Index Number:
207-330-6
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
88 °C/1013 hPa
vapor pressure:
70 hPa ( 20 °C)

$47.80


Check Cart for Availability

Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

70 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

174 °C

potency

3536 mg/kg LD50, oral (Rat)
>16600 mg/kg LD50, skin (Rat)

expl. lim.

1.5 % (v/v)

dilution

(for synthesis)

bp

88 °C/1013 hPa

mp

-66 °C

transition temp

flash point -6 °C

solubility

91 g/L

density

0.83 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(COCC)CC

InChI

1S/C5H12O2/c1-3-6-5-7-4-2/h3-5H2,1-2H3

InChI key

KLKFAASOGCDTDT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.829 - 0.832
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

23.0 °F - closed cup

Flash Point(C)

-5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service