Skip to Content
Merck
All Photos(1)

Key Documents

8.14464

Sigma-Aldrich

Gibberellic acid

for synthesis

Synonym(s):

Gibberellic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$45.20
1 G
$47.20
5 G
$166.00

$45.20


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$45.20
1 G
$47.20
5 G
$166.00

About This Item

Empirical Formula (Hill Notation):
C19H22O6
CAS Number:
Molecular Weight:
346.37
MDL number:
UNSPSC Code:
12352212
EC Index Number:
201-001-0
NACRES:
NA.22

$45.20


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥95.0% (HPLC)

form

solid

potency

6300 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

mp

233-235 °C

solubility

5 g/L

density

1.34 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1[C@]32[C@H]4[C@@]5([C@H]([C@@H]3C([C@H](C=C2)O)(C1=O)C)C(=O)O)C[C@@](CC4)(C(=C)C5)O

InChI

1S/C19H22O6/c1-9-7-17-8-18(9,24)5-3-10(17)19-6-4-11(20)16(2,15(23)25-19)13(19)12(17)14(21)22/h4,6,10-13,20,24H,1,3,5,7-8H2,2H3,(H,21,22)/t10-,11+,12-,13-,16?,17+,18+,19-/m1/s1

InChI key

IXORZMNAPKEEDV-QTWFBFKQSA-N

Application

  • Gibberellic Acid Can Improve Seed Germination and Ornamental Quality of Selected Cyclamen Species Grown Under Short and Long Days: This research shows that gibberellic acid significantly improves seed germination and ornamental qualities of cyclamen, suggesting applications in commercial floriculture for enhancing plant aesthetics and viability (Cornea-Cipcigan et al., 2020).
  • Synthesis of gibberellic acid derivatives and their effects on plant growth: This study explores the synthesis of novel gibberellic acid derivatives and tests their biological activity, providing a basis for the development of new agricultural chemicals aimed at enhancing crop yield and health (Tian et al., 2017).
  • Polymeric nanoparticles as an alternative for application of gibberellic acid in sustainable agriculture: The paper investigates the use of polymeric nanoparticles for delivering gibberellic acid to plants, aiming to increase its efficiency and reduce environmental impact, which could revolutionize practices in sustainable agriculture (Pereira et al., 2019).

Analysis Note

Assay (HPLC, area%): ≥ 95.0 % (a/a)
Spec. rotation [α²0/D (c=2 in methanol): +77.0 - +82.0 °
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service