Skip to Content
Merck
All Photos(1)

Key Documents

8.14444

Sigma-Aldrich

1-Octadecanethiol

for synthesis

Synonym(s):

1-Octadecanethiol, n-Octadecyl mercaptan, Stearyl mercaptane

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$41.10
50 G
$71.80
250 G
$276.00
8 KG
$3,360.00

$41.10


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
$41.10
50 G
$71.80
250 G
$276.00
8 KG
$3,360.00

About This Item

Linear Formula:
CH3-(CH2)16-CH2-SH
CAS Number:
Molecular Weight:
286.56
MDL number:
UNSPSC Code:
12352105
EC Index Number:
220-744-1
NACRES:
NA.22

$41.10


Estimated to ship on29 May 2025


Request a Bulk Order

vapor pressure

<0.5 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

solid

autoignition temp.

>200 °C

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

bp

366 °C

mp

28-31 °C

transition temp

flash point 185 °C

density

0.847 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

SCCCCCCCCCCCCCCCCCC

InChI

1S/C18H38S/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h19H,2-18H2,1H3

InChI key

QJAOYSPHSNGHNC-UHFFFAOYSA-N

Application

  • Self-Assembled 1-Octadecanethiol Membrane on Pd/ZnO for a Selective Room Temperature Flexible Hydrogen Sensor: This study explores the use of 1-octadecanethiol in the development of a selective room temperature hydrogen sensor, demonstrating its potential in enhancing sensor performance (Pathak, Cho, 2021).
  • Property improvement of GaAs surface by 1-octadecanethiol passivation: The article investigates the impact of 1-Octadecanethiol passivation on GaAs surfaces, revealing improvements in surface properties and potential applications in semiconductor technologies (Zhou et al., 2019).
  • Sputtering synthesis and optical investigation of octadecanethiol-protected fluorescent Au nanoparticles: This research examines the optical properties of gold nanoparticles protected by 1-octadecanethiol, highlighting their potential in various optical and electronic applications (Ishida et al., 2015).
  • Kinetics of radical telomerization of acrylic acid in the presence of 1-octadecanethiol: The study focuses on the telomerization process of acrylic acid with 1-octadecanethiol, offering insights into the polymerization kinetics and applications in polymer science (Mezhuev et al., 2018).
  • Formation of a mixed monolayer on a gold surface using fluorobenzenethiol and alkanethiol: This article describes the formation of mixed monolayers using 1-octadecanethiol, which could be crucial for developing advanced molecular assemblies on gold surfaces (Ikematsu et al., 2019).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Appearance of substance (visual): turbidity
Melting range (lower value): ≥ 28 °C
Melting range (upper value): ≤ 32 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service