Skip to Content
Merck
All Photos(1)

Key Documents

8.14283

Sigma-Aldrich

Bis(trichloromethyl) carbonate

for synthesis

Synonym(s):

Bis(trichloromethyl) carbonate, Triphosgene, Carbonic acid bistrichlor methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$65.40
25 G
$176.00
100 G
$210.00
1 KG
$380.00

$65.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$65.40
25 G
$176.00
100 G
$210.00
1 KG
$380.00

About This Item

Linear Formula:
(Cl3CO)2CO
CAS Number:
Molecular Weight:
296.75
MDL number:
UNSPSC Code:
12352108
EC Index Number:
250-986-3
NACRES:
NA.22

$65.40


Check Cart for Availability

Request a Bulk Order

vapor pressure

16 hPa ( 90 °C)

Quality Level

form

crystals

bp

203-206 °C/1013 mbar

mp

77-81 °C

density

1.6 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClC(Cl)(Cl)OC(=O)OC(Cl)(Cl)Cl

InChI

1S/C3Cl6O3/c4-2(5,6)11-1(10)12-3(7,8)9

InChI key

UCPYLLCMEDAXFR-UHFFFAOYSA-N

Application


  • Triphosgene in Organic Synthesis: Triphosgene is highlighted for its efficacy in organic reactions over the last decade, offering a safer and more convenient alternative to phosgene due to its solid state and lower volatility. so it can be used in various synthetic applications, enhancing reaction conditions and safety in laboratories(Ganiu et al., 2020).

  • Biodegradable Hydrogel Synthesis: Bis(trichloromethyl) carbonate was utilized in the one-step synthesis of biodegradable curcumin-derived hydrogels aimed at soft tissue filler applications post-breast cancer surgery. This represents a significant advancement in biomaterials, focusing on improving post-surgical recovery and compatibility (Shpaisman et al., 2012).

Analysis Note

Assay (ex Cl): ≥ 96.0 %
Melting range (lower value): ≥ 76 °C
Melting range (upper value): ≤ 81 °C
Identity (IR): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service