Skip to Content
Merck
All Photos(1)

Key Documents

8.14209

Sigma-Aldrich

Bis(4-hydroxyphenyl)sulfide

for synthesis

Synonym(s):

Bis(4-hydroxyphenyl)sulfide, 4,4′-Thiodiphenol, 4,4′-Dihydroxyphenyl sulfide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$41.40

$41.40


Estimated to ship on28 May 2025



Select a Size

Change View
100 G
$41.40

About This Item

Empirical Formula (Hill Notation):
C12H10O2S
CAS Number:
Molecular Weight:
218.27
MDL number:
UNSPSC Code:
12352104
EC Index Number:
220-197-9
NACRES:
NA.22

$41.40


Estimated to ship on28 May 2025


Quality Level

Assay

≥98.0% (GC)

potency

3362 mg/kg LD50, oral (Rat)
>10250 mg/kg LD50, skin (Rabbit)

mp

149-152 °C

storage temp.

2-30°C

SMILES string

S(c2ccc(cc2)O)c1ccc(cc1)O

InChI

1S/C12H10O2S/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12/h1-8,13-14H

InChI key

VWGKEVWFBOUAND-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 149 °C
Melting range (upper value): ≤ 153 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service