Skip to Content
Merck
All Photos(1)

Key Documents

8.14171

Sigma-Aldrich

Polyethylene glycol dimethyl ether 500

(stabilised) for synthesis

Synonym(s):

Polyethylene glycol dimethyl ether 500

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$124.00

$124.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
500 ML
$124.00

About This Item

Linear Formula:
CH3O(CH2CH2O)nCH3
CAS Number:
Molecular Weight:
530.65
MDL number:
UNSPSC Code:
12352112
NACRES:
NA.22

$124.00


Estimated to ship on27 May 2025


Request a Bulk Order

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

360 °C

potency

>2000 mg/kg LD50, oral (Rat)

kinematic viscosity

15 cSt(40 °C)

bp

>250 °C/1013 hPa

mp

13 °C

transition temp

flash point 220 °C

density

1.07 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCOC)C

InChI

1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3

InChI key

XTHFKEDIFFGKHM-UHFFFAOYSA-N

Application


  • Safety, high-performing and effects of the N/P ratio of a solid lithium ion battery using PEGDME based polymer electrolytes: This study examines the safety and performance characteristics of solid lithium ion batteries enhanced with Polyethylene glycol dimethyl ether (PEGDME) based polymer electrolytes, focusing on the N/P ratio′s impact (Jeon et al., 2023).

Analysis Note

Hydroxyl value: ≤ 2
pH-value (10 %; in water): 4.0 - 7.0
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

489.2 °F

Flash Point(C)

254 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service