Skip to Content
Merck
All Photos(1)

Key Documents

8.14119

Sigma-Aldrich

Hexadecyltrimethylammonium bromide

for synthesis

Synonym(s):

Hexadecyltrimethylammonium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$45.70
500 G
$80.00

$45.70


Available to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
$45.70
500 G
$80.00

About This Item

Linear Formula:
C16H33N(CH3)3Br
CAS Number:
Molecular Weight:
364.45
MDL number:
UNSPSC Code:
12352107
EC Index Number:
200-311-3
NACRES:
NA.22

$45.70


Available to ship on02 April 2025Details


Request a Bulk Order

Quality Level

form

solid

potency

410 mg/kg LD50, oral (Rat)

pH

5-7 (20 °C, 50 g/L in H2O)

mp

237-243 °C

solubility

55 g/L

bulk density

390 kg/m3

storage temp.

2-30°C

SMILES string

[Br-].[N+](CCCCCCCCCCCCCCCC)(C)(C)C

InChI

1S/C19H42N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H/q+1;/p-1

InChI key

LZZYPRNAOMGNLH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Development of porous particles: Utilizing Hexadecyltrimethylammonium bromide in microfluidics, researchers engineered interconnected open-porous particles, which could be applied in various industrial and pharmaceutical formulations (Masaoka et al., 2024).

Analysis Note

Assay (argentometric): ≥ 97.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Gastrointestinal tract, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

471.2 °F - closed cup

Flash Point(C)

244 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service