Skip to Content
Merck
All Photos(1)

Key Documents

8.14051

Sigma-Aldrich

Hexamethyldisiloxane

for synthesis

Synonym(s):

Hexamethyldisiloxane, 2,2,4,4-Tetramethyl-3-oxa-2,4-disilapentane, HMDSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3)3Si)2O
CAS Number:
Molecular Weight:
162.38
MDL number:
UNSPSC Code:
12352103
EC Index Number:
203-492-7
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

20 hPa ( 20 °C)

Quality Level

Assay

≥98.5% (GC)

form

liquid

autoignition temp.

340 °C

potency

>12160 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

expl. lim.

0.5-21.8 % (v/v)

bp

100 °C/1013 hPa

mp

-68 °C

transition temp

flash point 4 °C

solubility

insoluble 0.00037 g/L

density

0.76 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

[Si](O[Si](C)(C)C)(C)(C)C

InChI

1S/C6H18OSi2/c1-8(2,3)7-9(4,5)6/h1-6H3

InChI key

UQEAIHBTYFGYIE-UHFFFAOYSA-N

Application

  • Hexamethyldisiloxane coating by plasma to create a superhydrophobic surface for fabric masks: Research into the application of HMDSO plasma coatings to fabric masks to impart superhydrophobic properties, enhancing their resistance to moisture (Prado et al., 2022).
  • PECVD of hexamethyldisiloxane coatings using extremely asymmetric capacitive RF discharge: Examines the deposition of HMDSO-based coatings via plasma-enhanced chemical vapor deposition, focusing on the influence of discharge asymmetry on the coating properties (Gosar et al., 2020).
  • A ReaxFF-based molecular dynamics study of the pyrolysis mechanism of hexamethyldisiloxane: This study uses molecular dynamics to explore the pyrolysis processes of HMDSO, important for understanding its stability and decomposition under thermal stress (Chen et al., 2022).
  • Thermal stability of hexamethyldisiloxane and octamethyltrisiloxane: Discusses the thermal stability of HMDSO compared to related siloxanes, providing valuable data for applications involving heat exposure (Keulen et al., 2018).

Analysis Note

Assay (GC, area%): ≥ 98.5 % (a/a)
Density (d 20 °C/ 4 °C): 0.762 - 0.764
Identity (IR): passes test

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

21.2 °F - closed cup

Flash Point(C)

-6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service