This product is 70% (70 grams per 100 mL) tert-Butyl hydroperoxide in water. This refers to concentration.
8.14006
tert-Butyl hydroperoxide
(70% solution in water) for synthesis
Synonym(s):
1,1-Dimethylethyl hydroperoxide, 2-Hydroperoxy-2-methylpropane, TBHP
Select a Size
About This Item
synthesis grade
Recommended Products
grade
for synthesis
synthesis grade
Quality Level
vapor pressure
232 hPa ( 60 °C)
form
liquid
potency
810 mg/kg LD50, oral (Rat)
628 mg/kg LD50, skin (Rabbit)
reaction suitability
reagent type: oxidant
concentration
67.0-73.0% (iodometric)
dilution
(for synthesis)
refractive index
n20/D 1.3870
bp
37 °C/20 hPa
mp
-3 °C
transition temp
flash point 38 °C
solubility
130-150 g/L
density
0.94 g/cm3 at 20 °C
storage temp.
15-25°C
Analysis Note
Density (d 20 °C/ 4 °C): 0.937 - 0.938
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Org. Perox. F - Skin Corr. 1C - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
5.2 - Organic peroxides and self-reacting hazardous materials
WGK
WGK 3
Flash Point(F)
100.4 °F
Flash Point(C)
38 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
70% est ce une concentration massique ou molaire?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service