Skip to Content
Merck
All Photos(1)

Key Documents

8.11731

Sigma-Aldrich

Crown ether/Dibenzo-18-crown-6

for synthesis

Synonym(s):

Crown ether/Dibenzo-18-crown-6, Dibenzo-18-crown-6, 1,4,7,14,17,20-Hexaoxa[7.7]orthocyclophane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$67.40
50 G
$281.00

$67.40


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
$67.40
50 G
$281.00

About This Item

Empirical Formula (Hill Notation):
C20H24O6
CAS Number:
Molecular Weight:
360.40
MDL number:
UNSPSC Code:
12352005
EC Index Number:
238-041-3
NACRES:
NA.22

$67.40


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

powder

potency

2600 mg/kg LD50, oral (Rat)

mp

158-161 °C

storage temp.

2-30°C

SMILES string

O1CCOc2c(cccc2)OCCOCCOc3c(cccc3)OCC1

InChI

1S/C20H24O6/c1-2-6-18-17(5-1)23-13-9-21-11-15-25-19-7-3-4-8-20(19)26-16-12-22-10-14-24-18/h1-8H,9-16H2

InChI key

YSSSPARMOAYJTE-UHFFFAOYSA-N

Application

Crown ether/Dibenzo-18-crown-6 for synthesis. CAS 14187-32-7, molar mass 360.41 g/mol.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 158 °C
Melting range (upper value): ≤ 163 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service