810685
Probumin® Bovine Serum Albumin Media Grade
≥98%, USA origin, Tested for IgG, cell culture | mammalian: suitable, microbiological culture: suitable, Binding, transport studies
Synonym(s):
Albumin, BSA, Bovine Serum, Bovine Serum Albumin, Fraction V
Select a Size
About This Item
Recommended Products
Product Name
Probumin® Bovine Serum Albumin Media Grade, pkg of 1 kg
biological source
bovine serum
Quality Level
grade
media grade
Assay
≥98% (purity)
form
lyophilized powder
shelf life
48 mo.
mol wt
~66 kDa
purified by
heat shock fractionation
packaging
pkg of 1 kg
manufacturer/tradename
Probumin®
origin
USA origin
technique(s)
cell culture | mammalian: suitable
microbiological culture: suitable
dilution
(Binding, transport studies)
impurities
Tested for IgG
loss
≤5%
pH
6.8-7.2
anion traces
chloride (Cl-): 0-3.0 mg/g
cation traces
Na: 0-10 mg/g
foreign activity
Tested for Endotoxin
Tested for mycoplasma
Looking for similar products? Visit Product Comparison Guide
General description
Storage and Stability
Legal Information
Disclaimer
For further manufacturing use only. Not for human or animal consumption.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service