Skip to Content
Merck
All Photos(2)

Key Documents

8.08505

Sigma-Aldrich

Vanadium(IV) oxide acetylacetonate

for synthesis

Synonym(s):

Vanadium(IV) oxide acetylacetonate, Bis(acetylacetonato)vanadium(IV) oxide, Bis(2,4-pentanedionato)vanadium(IV) oxide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$163.00

$163.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
$163.00

About This Item

Empirical Formula (Hill Notation):
C10H14O5V
CAS Number:
Molecular Weight:
265.16
MDL number:
UNSPSC Code:
12352103
EC Index Number:
221-590-8
NACRES:
NA.22

$163.00


Available to ship on31 March 2025Details


Request a Bulk Order

Quality Level

form

powder

mp

243-246 °C

solubility

10 g/L

bulk density

450 kg/m3

storage temp.

2-30°C

SMILES string

[V]=O.O=C(CC(=O)C)C.O=C(CC(=O)C)C

InChI

1S/2C5H8O2.O.V/c2*1-4(6)3-5(2)7;;/h2*3H2,1-2H3;;

InChI key

GGUKAPURIDGXAJ-UHFFFAOYSA-N

Analysis Note

Assay (ex V): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service