Skip to Content
Merck
All Photos(1)

Key Documents

8.08332

Sigma-Aldrich

Ethyl 4-toluenesulfonate

for synthesis

Synonym(s):

Ethyl 4-toluenesulfonate, p-Toluenesulfonic acid ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$59.60
500 G
$180.00
5 KG
$772.00

$59.60


0 left in stock

Request a Bulk Order

Select a Size

Change View
100 G
$59.60
500 G
$180.00
5 KG
$772.00

About This Item

Linear Formula:
4-(CH3)C6H4SO3C2H5
CAS Number:
Molecular Weight:
200.25
MDL number:
UNSPSC Code:
12352108
EC Index Number:
201-276-7
NACRES:
NA.22

$59.60


0 left in stock

Request a Bulk Order

vapor pressure

0.6 hPa ( 14 °C)

Quality Level

Assay

≥97.0% (GC)

form

solid

autoignition temp.

500 °C

reaction suitability

reagent type: reductant

bp

221 °C/1013 hPa

mp

30-32 °C

transition temp

flash point 158 °C

density

1.174 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(OCC)c1ccc(cc1)C

InChI

1S/C9H12O3S/c1-3-12-13(10,11)9-6-4-8(2)5-7-9/h4-7H,3H2,1-2H3

InChI key

VRZVPALEJCLXPR-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 29 °C
Melting range (upper value): ≤ 32 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

316.4 °F - closed cup

Flash Point(C)

158 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service